EMAIL THIS PAGE TO A FRIEND
Radiatsionnaia biologiia, radioecologiia
[The antioxidant enzyme activity in mouse liver mitochondria after nanosecond pulsed periodic X-ray exposure].
PMID 23786032
Abstract
The effect of repetitive pulsed X-ray (4 ns pulse duration, 300 kV accelerating voltage; 2.5 kA electron beam current) on the antioxidant enzyme activity in mouse liver mitochondria has been investigated. The mitochondrial suspension was exposed to single 4000 pulse X-ray radiation with repetition rates ranging between 10 and 22 pps (pulsed dose was 0.3-1.8 x 10(-6) Gy/pulse, the total absorbed dose following a single exposure was 7.2 x 10(-3) Gy). It was shown that a short-time exposure to X-ray radiation changes the antioxidant enzyme activity in mouse liver mitochondria. The greatest effect was observed in the changes of the activity of the metal-containing enzymes: superoxide dismutase and glutathione peroxidase. The effect depends on the pulse repetition frequency and radiation dose.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C3515 Catalase from Aspergillus niger, ammonium sulfate suspension, ≥4,000 units/mg protein
C40 Catalase from bovine liver, lyophilized powder, ≥10,000 units/mg protein
C30 Catalase from bovine liver, aqueous suspension, 10,000-40,000 units/mg protein
C100 Catalase from bovine liver, aqueous suspension, 40,000-60,000 units/mg protein (E1%/405)
C3155 Catalase from bovine liver, aqueous solution, ≥30,000 units/mg protein
02071 Catalase from Corynebacterium glutamicum, solution, deep brown, ≥500000 U/mL
C3556 Catalase from human erythrocytes, ≥90% (SDS-PAGE), buffered aqueous solution, ≥30,000 units/mg protein
C9284 Catalase−Agarose, pH 5.4(isoelectric point), ammonium sulfate suspension, enzyme from bovine liver
G6137 Glutathione Peroxidase from bovine erythrocytes, lyophilized powder, ≥300 units/mg protein
S9697
Superoxide Dismutase bovine, recombinant, expressed in E. coli, lyophilized powder, ≥2500 units/mg protein, ≥90% (SDS-PAGE)
S7571
Superoxide Dismutase from bovine erythrocytes, lyophilized powder, ≥3,000 units/mg protein
S5395
Superoxide Dismutase from bovine erythrocytes, ≥3,000 units/mg protein, BioReagent, lyophilized powder, suitable for cell culture
S7446
Superoxide Dismutase from bovine erythrocytes, BioUltra, lyophilized powder, ≥4,500 units/mg protein, ≥97% (SDS-PAGE)
S8160
Superoxide Dismutase from bovine liver, lyophilized powder, ≥1500 units/mg protein
S8409
Superoxide Dismutase from bovine liver, ammonium sulfate suspension, 2,000-6,000 units/mg protein (biuret)
S5639
Superoxide Dismutase from Escherichia coli, lyophilized powder, ≥1,000 units/mg protein
S9636
Superoxide Dismutase from human erythrocytes, essentially salt-free, lyophilized powder, ≥2,500 units/mg protein
S9076 Superoxide Dismutase I human, recombinant, expressed in E. coli