EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Gold-catalyzed cyclizations of cis-enediynes: insights into the nature of gold-aryne interactions.
PMID 23788447
Abstract
Golden aryne? Gold aryne complexes are inferred as transition states in dual gold-catalyzed cyclizations of cis-enediynes (see scheme; DCE = 1,2-dichloroethane). They are better described as ortho-aurophenyl cations, which react with weak nucleophiles and undergo facile intramolecular insertions into C(sp(3))-H bonds. Indanes, fused heteroarenes, and phenol derivatives are readily prepared using this method.