EMAIL THIS PAGE TO A FRIEND
The Journal of Biological Chemistry
Peroxisome proliferator-activated receptor ? (PPAR?) and its target genes are downstream effectors of FoxO1 protein in islet ?-cells: mechanism of ?-cell compensation and failure.
PMID 23788637
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AB4130 Anti-FOXO1 Antibody, clone, Chemicon®, from rabbit