EMAIL THIS PAGE TO A FRIEND
Anesthesia and analgesia
The effective effect-site propofol concentration for induction in morbidly obese patients using total body weight with a new pharmacokinetic model.
PMID 23788741