EMAIL THIS PAGE TO A FRIEND
Organic preparations and procedures international
Comparative Study of the Frech Catalyst with Two Conventional Catalysts in the Heck Synthesis of 2,4-Diaminopyrimidine-based Antibiotics.
PMID 23788820