EMAIL THIS PAGE TO A FRIEND
Sudebno-meditsinskaia ekspertiza
[The distribution of 4-nitroanilin in the body of warm-blooded animals].
PMID 23789413
Abstract
Specific distribution patterns of 4-nitroanilin in the body of warm-blooded animals (rats) were studied after the in vivo administration of this poisonous substance. It was shown that unmetabolized 4-nitroanilin accumulates in large quantities in the internal organs of the intoxicated animals. Its highest levels are found in the stomach and its contents, kidneys, and lungs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
185310
4-Nitroaniline, ≥99%
C6H6N2O2
31569
4-Nitroaniline, analytical standard
C6H6N2O2