EMAIL THIS PAGE TO A FRIEND
Undersea & hyperbaric medicine : journal of the Undersea and Hyperbaric Medical Society, Inc
Edaravone may be an efficacious adjunctive therapeutic agent for decompression sickness: an editorial perspective.
PMID 23789554
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M70800
3-Methyl-1-phenyl-2-pyrazoline-5-one, 99%
C10H10N2O