EMAIL THIS PAGE TO A FRIEND
Undersea & hyperbaric medicine : journal of the Undersea and Hyperbaric Medical Society, Inc
The free radical scavenger, edaravone, ameliorates delayed neuropsychological sequelae after acute carbon monoxide poisoning in rabbits.
PMID 23789557
Abstract
The mechanism underlying delayed neuropsychological sequelae (DNS) after acute carbon monoxide (CO) poisoning is unclear. There are no effective treatments for DNS. As part of a new generation of antioxidants, edaravone has been reported to improve clinical outcomes in patients exhibiting ischemic strokes. There has been little data about edaravone in relationship to DNS prevention and treatment. We hypothesized that edaravone could ameliorate DNS: Here we test that hypothesis in rabbits Rabbits were randomly divided into sham control,DNS group, saline group and edaravone group. DNS model was made by intraperitoneal injection of CO. Normal saline or edaravone (1 mg/kg, twice daily, a total of one course for 14 days) was infused through the ear vein from Day 15 since the DNS model was established. Serum superoxide dismutase (SOD) activity and malondialdehyde (MDA) were measured in each group. Magnetic resonance spectroscopy (MRS) was used to examine regions of the brain for various compounds. The apoptotic index and neuronal density in the hippocampal CA1 area were also investigated. SOD activity decreased significantly and MDA content increased substantially in the DNS group and saline group when compared with the sham control (p < 0.01). Conversely, in the edaravone group, serum SOD activity significantly increased and MDA levels significantly decreased when compared with DNS and saline group (p < 0.01). In the DNS group, the spectra of H1-MRS showed an elevated Cho/Cr and Lac/Cr ratio, and a marked decrease in the NAA/Cr ratio (p < 0.01). Compared with the saline group and DNS group, the NAA/Cr ratio was significantly increased, and the Cho/Cr and Lac/Cr ratio were significantly decreased in the edaravone group (p < 0.01). The apoptotic index in the edaravone group was significantly lower than that of the DNS and saline groups (p < 0.01, respectively), while the neuronal density in edaravone group was significantly higher than that of the DNS and saline group in the hippocampal CA1 area (p < 0.01, respectively). Our present research demonstrates that edaravone could ameliorate DNS after acute carbon monoxide poisoning in rabbits. These results suggest free radicals could be involved in the underlying mechanisms of DNS. Furthermore, brain MRS shows promise as a tool for early diagnosis for DNS.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
68740
3-Methyl-1-phenyl-2-pyrazoline-5-one, purum, ≥98.0% (NT)
C10H10N2O
M70800
3-Methyl-1-phenyl-2-pyrazoline-5-one, 99%
C10H10N2O
10784
Antipyrine, tested according to Ph.Eur.
C11H12N2O
10790
Antipyrine, ≥99.0% (RT)
C11H12N2O
11260
L-Aspartic acid hemimagnesium salt dihydrate, ≥98.0% (KT)
C8H12MgN2O8 · 2H2O
S9697
Superoxide Dismutase bovine, recombinant, expressed in E. coli, lyophilized powder, ≥2500 units/mg protein, ≥90% (SDS-PAGE)
S2515
Superoxide Dismutase from bovine erythrocytes, lyophilized powder, 2,500-7,000 units/mg protein
S5395
Superoxide Dismutase from bovine erythrocytes, BioReagent, ≥3,000 units/mg protein, suitable for cell culture, lyophilized powder
S7571
Superoxide Dismutase from bovine erythrocytes, lyophilized powder, ≥3,000 units/mg protein
S7446
Superoxide Dismutase from bovine erythrocytes, BioUltra, lyophilized powder, ≥4,500 units/mg protein, ≥97% (SDS-PAGE)
S8160
Superoxide Dismutase from bovine liver, lyophilized powder, ≥1500 units/mg protein
S8409
Superoxide Dismutase from bovine liver, ammonium sulfate suspension, 2,000-6,000 units/mg protein (biuret)
S5639
Superoxide Dismutase from Escherichia coli, lyophilized powder, ≥1,000 units/mg protein
S5389
Superoxide Dismutase from Escherichia coli, lyophilized powder, ≥1,000 units/mg protein
S9636
Superoxide Dismutase from human erythrocytes, essentially salt-free, lyophilized powder, ≥2,500 units/mg protein
S9076 Superoxide Dismutase I human, recombinant, expressed in E. coli