EMAIL THIS PAGE TO A FRIEND
American journal of obstetrics and gynecology
Superoxide dismutase 1 overexpression in mice abolishes maternal diabetes-induced endoplasmic reticulum stress in diabetic embryopathy.
PMID 23791840
Abstract
Both oxidative stress and endoplasmic reticulum stress (ER stress) are causal events in diabetic embryopathy. We tested whether oxidative stress causes ER stress. Wild-type (WT) and superoxide dismutase 1 (SOD1)-overexpressing day 8.75 embryos from nondiabetic WT control with SOD1 transgenic male and diabetic WT female with SOD1 transgenic male were analyzed for ER stress markers: C/EBP-homologous protein (CHOP), calnexin, eukaryotic initiation factor 2α (eIF2α), protein kinase ribonucleic acid (RNA)-like ER kinase (PERK), binding immunoglobulin protein, protein disulfide isomerase family A member 3, kinases inositol-requiring protein-1α (IRE1α), and the X-box binding protein (XBP1) messenger RNA (mRNA) splicing. Maternal diabetes significantly increased the levels of CHOP, calnexin, phosphorylated (p)-eIF2α, p-PERK, and p-IRE1α; triggered XBP1 mRNA splicing; and enhanced ER chaperone gene expression in WT embryos. SOD1 overexpression blocked these diabetes-induced ER stress markers. Mitigating oxidative stress via SOD1 overexpression blocks maternal diabetes-induced ER stress in vivo.