EMAIL THIS PAGE TO A FRIEND
Technology and health care : official journal of the European Society for Engineering and Medicine
Refractory adhesive capsulitis under acitretin therapy.
PMID 23792799
Abstract
Adhesive capsulitis (AC) is characterized by a limited active and passive motion. Although the exact pathology remains unknown, a number of contributing factors are discussed. AC has probably been caused by the Re-PUVA therapy (PUVA irradiation plus acitretin) of a cutaneous T-cell lymphoma, type mycosis fungoides. Acitretin belongs to the group of retinoids and is often used in cornification disorders. After non-successful initial conservative therapy with intraarticular steroid injections and physical therapy, a significant improvement of shoulder joint mobility was finally achieved by an arthroscopic juxtaglenoid capsulotomy and adhesiolysis. A therapy with acitretin should be considered as a possible trigger of AC. Patient's medication should be checked carefully on possible triggers of AC. The athroscopic adhesiolysis is an effective method for a frustrating conservative treatment of AC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1011018 Acitretin Related Compound A, United States Pharmacopeia (USP) Reference Standard
C21H26O3