EMAIL THIS PAGE TO A FRIEND
Zhongguo gu shang = China journal of orthopaedics and traumatology
[Effects of geniposide on SNP-induced apoptosis of chondrocyte and cell cycle].
PMID 23795444
Abstract
To study the effects of Geniposide on SNP(sodium nitroprusside)-induced apoptosis of chondrocyte in vitro and cell cycle. The chondrocyte of three-week-old SD rats were separated and cultivated. The second generation of chondrocyte cells were involved in experiment. Chondrocyte proliferation was measured by assay; flow cytometer were adopted to observe cell cycle and apoptosis rate; NO examination adopted nitrate reductase method. Geniposide could significantly decrease the percentage of SNP-induced chondrocytes in G0/G1 phase and increased percentage in S phase and G2/M phase. The apoptosis of chondrocyte and the concentration of NO in the culture supernatants was reduced significantly (r=0.917, P<0.01). Geniposide could impact SNP-induced apoptosis of chondrocyte by reducing the concentration of NO in the culture supernatants, promoting proliferation of chondrocytes, which is a probable and important mechanism of Geniposide preventing osteoarthritis.