EMAIL THIS PAGE TO A FRIEND
Biochemical Society transactions
Dexamethasone and phenylalanine hydroxylase activities in rats.
PMID 2379736
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D0387
6,7-Dimethyl-5,6,7,8-tetrahydropterine hydrochloride, ≥95%
C8H13N5O · HCl