EMAIL THIS PAGE TO A FRIEND
Circulation
Letter by Ruggieri et al regarding article, "Randomized comparison of sevoflurane versus propofol to reduce perioperative myocardial ischemia in patients undergoing noncardiac surgery".
PMID 23797747