EMAIL THIS PAGE TO A FRIEND
The Medical letter on drugs and therapeutics
Tobramycin inhalation powder (Tobi Podhaler) for cystic fibrosis.
PMID 23797798
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0001210
Tobramycin for identification, European Pharmacopoeia (EP) Reference Standard
C18H37N5O9