EMAIL THIS PAGE TO A FRIEND
Environmental and molecular mutagenesis
Microsomal epoxide hydrolase (EPHX1) polymorphisms are associated with aberrant promoter methylation of ERCC3 and hematotoxicity in benzene-exposed workers.
PMID 23797950
Abstract
Benzene is an important industrial chemical and widespread environmental pollutant known to induce leukemia and other blood disorders. To be carcinogenic, benzene must be metabolized to produce toxic metabolites. To investigate whether single nucleotide polymorphisms (SNPs) in the metabolic enzyme genes are associated with benzene-induced alterations in DNA methylation and hematotoxicity, we genotyped four commonly studied SNPs in three metabolic enzymes genes CYP1A1, EPHX1 and NQO1; and analyzed promoter DNA methylation status in 11 genes which have been reported to be associated with benzene-induced hematotoxicity (BLM, CYP1A1, EPHX1, ERCC3, NQO1, NUDT1, p15, p16, RAD51, TP53 and WRAP53) in 77 benzene-exposed workers and 25 unexposed controls in China. ERCC3, a DNA repair gene, showed a small but statistically significant increase of promoter DNA methylation in the exposed group compared with the unexposed group (mean ± SD: 4.73 ± 3.46% vs. 3.63 ± 1.96%, P = 0.048). We also observed that an increased number of C allele for rs1051740 in EPHX1 was associated with decreased ERCC3 methylation levels in benzene-exposed workers (P(trend)  = 0.001), but not in unexposed controls (P(trend)  = 0.379). Interestingly, another EPHX1 SNP (rs2234922) was associated with lower white blood cell (WBC) counts (P(trend)  = 0.044) in benzene-exposed workers. These associations remained the same when ERCC3 promoter methylation and WBCs were dichotomized according to the 90th percentile (≥6%) of methylation levels in controls and a leucopenia cutoff (<4 × 10(9) /L), respectively. Our findings suggest that benzene exposure may be associated with hypermethylation in ERCC3, and that genetic variants in EPHX1 may play an important role in epigenetic changes and hematotoxicity among benzene-exposed workers.