PloS one
Synthesis of coral-like tantalum oxide films via anodization in mixed organic-inorganic electrolytes.
PMID 23799106
Abstract
We report a simple method to fabricate nano-porous tantalum oxide films via anodization with Ta foils as the anode at room temperature. A mixture of ethylene glycol, phosphoric acid, NH4F and H2O was used as the electrolyte where the nano-porous tantalum oxide could be synthesized by anodizing a tantalum foil for 1 h at 20 V in a two-electrode configuration. The as-prepared porous film exhibited a continuous, uniform and coral-like morphology. The diameters of pores ranged from 30 nm to 50 nm. The pores interlaced each other and the depth was about 150 nm. After calcination, the as-synthesized amorphous tantalum oxide could be crystallized to the orthorhombic crystal system. As observed in photocatalytic experiments, the coral-like tantalum oxide exhibited a higher photocatalytic activity for the degradation of phenol than that with a compact surface morphology, and the elimination rate of phenol increased by 66.7%.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF34465888
Tantalum, foil, 0.5m coil, thickness 0.034mm, as rolled, 99.9%
Ta
GF96084603
Tantalum, foil, 0.5m coil, thickness 0.03mm, as rolled, 99.9%
Ta
GF78648092
Tantalum, foil, 0.5m coil, thickness 0.03mm, coil width 1.2mm, annealed, 99.9%
Ta
GF38365106
Tantalum, foil, 0.5m coil, thickness 0.04mm, coil width .65mm, as rolled, 99.9%
Ta
GF76540181
Tantalum, foil, 0.5m coil, thickness 0.04mm, coil width 0.7mm, annealed, 99.9%
Ta
GF08928108
Tantalum, foil, 0.5m coil, thickness 0.04mm, coil width 1mm, annealed, 99.9%
Ta
GF61023574
Tantalum, foil, 0.5m coil, thickness 0.05mm, annealed, 99.9%
Ta
GF07210347
Tantalum, foil, 10m coil, thickness 0.025mm, coil width 0.75mm, annealed, 99.9%
Ta
GF10417698
Tantalum, foil, 10m coil, thickness 0.03mm, coil width 0.75mm, annealed, 99.9%
Ta
GF53229269
Tantalum, foil, 10m coil, thickness 0.04mm, coil width .65mm, as rolled, 99.9%
Ta
GF76534500
Tantalum, foil, 100x100mm, thickness 0.100mm, annealed, 99.9%
Ta
GF80276182
Tantalum, foil, 100x100mm, thickness 0.125mm, annealed, 99.9%
Ta
GF85784653
Tantalum, foil, 100x100mm, thickness 0.15mm, annealed, 99.9%
Ta
GF65422739
Tantalum, foil, 100x100mm, thickness 0.15mm, annealed, flattened, 99.9%
Ta
GF86342485
Tantalum, foil, 100x100mm, thickness 0.20mm, annealed, 99.9%
Ta
GF60298195
Tantalum, foil, 100x100mm, thickness 0.25mm, annealed, 99.9%
Ta
GF32441543
Tantalum, foil, 100x100mm, thickness 0.25mm, annealed, 99.99%
Ta
GF48780142
Tantalum, foil, 100x100mm, thickness 0.5mm, annealed, 99.9%
Ta
GF66145260
Tantalum, foil, 100x100mm, thickness 0.5mm, annealed, polished on both sides, 99.9%
Ta
GF55289548
Tantalum, foil, 15mm disks, thickness 0.5mm, annealed, 99.9%
Ta
GF64539872
Tantalum, foil, 15mm disks, thickness 0.75mm, annealed, 99.9%
Ta
GF24066525
Tantalum, foil, 1m coil, thickness 0.006mm, 99.9%
Ta
GF50723243
Tantalum, foil, 1m coil, thickness 0.009mm, 99.9%
Ta
GF45003204
Tantalum, foil, 1m coil, thickness 0.015mm, 99.9%
Ta
GF94153754
Tantalum, foil, 1m coil, thickness 0.025mm, as rolled, 99.9%
Ta
GF19196310
Tantalum, foil, 1m coil, thickness 0.025mm, coil width 0.75mm, annealed, 99.9%
Ta
GF13236487
Tantalum, foil, 1m coil, thickness 0.03mm, coil width 0.75mm, annealed, 99.9%
Ta
GF53856186
Tantalum, foil, 1m coil, thickness 0.03mm, coil width 1.2mm, annealed, 99.9%
Ta
GF10672365
Tantalum, foil, 1m coil, thickness 0.04mm, coil width .65mm, as rolled, 99.9%
Ta
GF67523351
Tantalum, foil, 1m coil, thickness 0.04mm, coil width 0.7mm, annealed, 99.9%
Ta