EMAIL THIS PAGE TO A FRIEND
Journal of clinical microbiology
Antibody to cytotoxin in infection by Helicobacter pylori.
PMID 2380350
Abstract
Gastrointestinal disease and colonization by Helicobacter pylori were determined in 36 asymptomatic volunteers and 30 symptomatic individuals undergoing endoscopy and biopsy. Serum antibody immunoglobulin G (IgG) and IgA to H. pylori were measured by enzyme-linked immunosorbent assay. Serum antibody to a cytotoxin produced by H. pylori was detected with a neutralization assay. Serum IgG was 95% predictive of infection by H. pylori, and serum IgA was 88% predictive. Antibody to the cytotoxin was detected in 12 of 18 infected individuals. Antibody to the cytotoxin was a highly specific (96%), but not a very sensitive (67%), indicator of infection by H. pylori. The neutralization assay was 87% predictive of infection. These data confirm the diagnostic value of serum antibody to H. pylori for the detection of infection. The toxin-neutralizing activity of sera from individuals infected with H. pylori suggests that the cytotoxin is produced in vivo. It may therefore contribute to disease associated with H. pylori.