EMAIL THIS PAGE TO A FRIEND
Environmental toxicology
Evaluation of cytotoxic, oxidative stress, proinflammatory and genotoxic effect of silver nanoparticles in human lung epithelial cells.
PMID 23804405
Abstract
Silver nanoparticles are increasingly used in various products, due to their antibacterial properties. Despite its wide spread use, only little information on possible adverse health effects exists. Therefore, the aim of this study was to assess the toxic potential of silver nanoparticles (<100 nm) in human lung epithelial (A549) cells and the underlying mechanism of its cellular toxicity. Silver nanoparticles induced dose and time-dependent cytotoxicity in A549 cells demonstrated by MTT and LDH assays. Silver nanoparticles were also found to induce oxidative stress in dose and time-dependent manner indicated by depletion of GSH and induction of ROS, LPO, SOD, and catalase. Further, the activities of caspases and the level of proinflammatory cytokines, namely interleukin-1β (IL-1β) and interleukin-6 (IL-6) were significantly higher in treated cells. DNA damage, as measured by single cell gel electrophoresis, was also dose and time-dependent signicants in A549 cells. This study investigating the effects of silver nanoparticles in human lung epithelial cells has provided valuable insights into the mechanism of potential toxicity induced by silver nanoparticles and warrants more careful assessment of silver nanoparticles before their industrial applications.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T3251
(±)-α-Tocopherol, synthetic, ≥96% (HPLC)
C29H50O2
90669
(±)-α-Tocopherol, tested according to Ph Eur
C29H50O2
47783
(±)-α-Tocopherol, analytical standard
C29H50O2
T3634
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
C29H50O2
T1539
(+)-α-Tocopherol, Type VI, from vegetable oil, neat (liquid, ≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
C29H50O2
127698
2-Nitrobenzoic acid, 95%, Contains 3- and 4-isomers
C7H5NO4
T1550000
α-Tocopherol, European Pharmacopoeia (EP) Reference Standard
C29H50O2
258024
α-Tocopherol, ≥95.5%
C29H50O2
1009005
Acetylcysteine, United States Pharmacopeia (USP) Reference Standard
C5H9NO3S
A0150000
Acetylcysteine, European Pharmacopoeia (EP) Reference Standard
C5H9NO3S
A5508
Adenosine 5′-phosphosulfate sodium salt, ≥85%
C10H14N5O10PS
PHR1031
Alpha Tocopherol, Pharmaceutical Secondary Standard; Certified Reference Material
C29H50O2
1667600
Alpha Tocopherol, United States Pharmacopeia (USP) Reference Standard
C29H50O2
A3678
Ammonium persulfate, for molecular biology, for electrophoresis, ≥98%
H8N2O8S2
A7460
Ammonium persulfate, BioXtra, ≥98.0%
H8N2O8S2
09913
Ammonium persulfate, BioUltra, for molecular biology, ≥98.0% (RT)
H8N2O8S2
PHR1098
N-Acetyl-L-cysteine, Pharmaceutical Secondary Standard; Certified Reference Material
C5H9NO3S
223441
Sodium hydride, dry, 90%
HNa
452912
Sodium hydride, 60 % dispersion in mineral oil
HNa
71620
Sodium hydride, powder (moistened with oil), 55-65% (gas-volumetric)
HNa