EMAIL THIS PAGE TO A FRIEND
Akusherstvo i ginekologiia
[Intrahepatic cholestasis of pregnancy].
PMID 23805459
Abstract
The contemporary aspects of obstetric cholestasis including its etiology, pathogenesis, molecular basis, diagnosis, therapy and obstetric management are presented. The possible maternal and fetal complications are discussed too.