EMAIL THIS PAGE TO A FRIEND
Journal of zoo and wildlife medicine : official publication of the American Association of Zoo Veterinarians
Successfully treated dermatomycosis in California sea lions (Zalophus californianus).
PMID 23805567
Abstract
We describe clinical cases caused by Microsporum gypseum in two subadult male California sea lions (Zalophus californianus). Dermatomycosis is uncommonly reported in pinnipeds, including this species. In these cases, skin lesions were multifocal to coalescing, involved all flippers, and were most pronounced on the ventral surfaces of flippers. They were well-demarcated, depigmented, and covered with crusts. The definitive diagnosis was obtained through microscopic examination and fungal culture of skin scrapings. Oral terbinafine and topical enilconazole were used as treatments for 65 days, and complete recovery was subsequently achieved. California sea lion, dermatomycosis, Microsporum gypseum, terbinafine, enilconazole
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32007
Imazalil, PESTANAL®, analytical standard
C14H14Cl2N2O
33997
Imazalil sulfate, PESTANAL®, analytical standard
C14H14Cl2N2O · H2SO4