EMAIL THIS PAGE TO A FRIEND
Patologicheskaia fiziologiia i eksperimental'naia terapiia
[Direct involvement of the glucose and Mg(2+)-ATP in the regulation of the GABAA-coupled Cl-,H(CO3(-) -activated Mg(2+)-ATPase of the plasma membrane from rat brain in vitro experiences].
PMID 23805711
Abstract
Effect of the glucose and Mg(2+)-ATP on the coupled with CABA(A)-receptor Cl-,HCO3(-)-activated Mg(2+)-ATPase of the plasma membranes from rat brain involving from "basal" Mg(2+)-ATPase which it is activated by Cl-,HCO3(-) ions it was investigated. The glucose (1-10 mM) decreased the "basal" Mg(2+)-ATPase activity on 17% and completely eliminated the enzyme activation by 10 mM Cl(-)+2 mM HCO3(-) ions. The variety effect of the glucose and Mg(2+)-ATP on the activation of the '"basal" Mg(2+)-ATPase by variety concentrations of anions it was established. So it was found that in the presence Mg(2+)-ATP in the incubation medium > 1 mM the enzyme activation by 10 mM Cl- + 2 mM HCO ions not appear. However, in the presence of the glucose (10 mM) the inhibiting effect of the Mg(2+)-ATP on the enzyme is disappears and Cl-,HCO3(-)-ATPase activity is restored. While, in the presence of the high concentrations 40 mM Cl- + 8 mM HCO3(-) in the incubation medium the inhibiting effect of the glucose and Mg(2+)-ATP it was negligible (-20%). It was conclusion about direct involvement of the glucose and Mg(2+)-ATP in the regulation of the coupled with CABA(A)-receptor Cl-,HCO3(-)-ATPase activity of the neuronal membrane under different concentration anions in the incubation medium.