EMAIL THIS PAGE TO A FRIEND
Heart rhythm
Kv7.1 isoform gradients in the heart: new potential approach to alter repolarization reserve.
PMID 23806322
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1019701 Amiloride hydrochloride, United States Pharmacopeia (USP) Reference Standard
C6H8ClN7O · HCl · 2H2O