EMAIL THIS PAGE TO A FRIEND
Annals of allergy, asthma & immunology : official publication of the American College of Allergy, Asthma, & Immunology
Eosinophilic esophagitis to unsuspected rare food allergen.
PMID 23806462