EMAIL THIS PAGE TO A FRIEND
Annals of allergy, asthma & immunology : official publication of the American College of Allergy, Asthma, & Immunology
Rapid desensitization to doxycycline.
PMID 23806469
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D3072 Doxycycline Hydrochloride, Ready Made Solution
C22H24N2O8 · HCl