EMAIL THIS PAGE TO A FRIEND
Eye (London, England)
Are generic topical prostanoids the way forward in the care of glaucoma patients? - Yes.
PMID 23807388
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6116
(+/-)-Cloprostenol sodium salt hydrate, ≥98% (HPLC), powder
C22H28ClNaO6 · xH2O