EMAIL THIS PAGE TO A FRIEND
Mikrobiologiia
[The influence of bacterial autoregulation molecules (homoserine lactones and alkyloxybenzoles) on oxidative metabolism of the natural immunity cellular effectors].
PMID 23808138