EMAIL THIS PAGE TO A FRIEND
Molekuliarnaia biologiia
[G4-quadruplexes and genome instability].
PMID 23808155
Abstract
A comparative analysis in silico of distribution of nucleotide sequences that predispose to formation of non-canonical DNA structure of G-quadruplexes, closely related with gene expression regulation and double strand DNA breaks, within vertebrata and yeast nuclear and mitochondrial genomes was carried out. Data on preferable localization of potential quadruplexes within non-coding sequences, their evolutionary conservation, and existing homology between them in mitochondrial and nuclear genomes were obtained. A possible interrelation between quadruplexes, Pif1 helicase and genomic instability is discussed.