EMAIL THIS PAGE TO A FRIEND
Anaesthesia and intensive care
Dobutamine induced cardiogenic shock due to systolic anterior motion after mitral valve repair.
PMID 23808519
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D2954000
Dobutamine hydrochloride, European Pharmacopoeia (EP) Reference Standard
C18H23NO3 · HCl