EMAIL THIS PAGE TO A FRIEND
Molecular immunology
Complement therapy in atypical haemolytic uraemic syndrome (aHUS).
PMID 23810412
Abstract
Central to the pathogenesis of atypical haemolytic uraemic syndrome (aHUS) is over-activation of the alternative pathway of complement. Inherited defects in complement genes and autoantibodies against complement regulatory proteins have been described. The use of plasma exchange to replace non-functioning complement regulators and hyper-functional complement components in addition to the removal of CFH-autoantibodies made this the 'gold-standard' for management of aHUS. In the last 4 years the introduction of the complement inhibitor Eculizumab has revolutionised the management of aHUS. In this review we shall discuss the available literature on treatment strategies to date.