EMAIL THIS PAGE TO A FRIEND
Environmental pollution (Barking, Essex : 1987)
Influence of biochar on isoproturon partitioning and bioaccessibility in soil.
PMID 23810820
Abstract
The influence of biochar (5%) on the loss, partitioning and bioaccessibility of (14)C-isoproturon ((14)C-IPU) was evaluated. Results indicated that biochar had a dramatic effect upon (14)C-IPU partitioning: (14)C-IPU extractability (0.01 M CaCl2) in biochar-amended treatments was reduced to <2% while, (14)C-IPU extractability in biochar free treatments decreased with ageing from 90% to 40%. A partitioning model was constructed to derive an effective partition coefficient for biochar:water (KBW of 7.82 × 10(4) L kg(-1)). This was two orders of magnitude greater than the apparent Kfoc value of the soil organic carbon:water (631 L kg(-1)). (14)C-radiorespirometry assays indicated high competence of microorganisms to mineralise (14)C-IPU in the absence of biochar (40.3 ± 0.9%). Where biochar was present (14)C-IPU mineralisation never exceeded 2%. These results indicate reduced herbicide bioaccessibility. Increasing IPU application to ×10 its recommended dose was ineffective at redressing IPU sequestration and its low bioaccessibility.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36137
Isoproturon, PESTANAL®, analytical standard
C12H18N2O