EMAIL THIS PAGE TO A FRIEND
Pakistan journal of pharmaceutical sciences
Comparison of chemical composition of Aerva javanica seed essential oils obtained by different extraction methods.
PMID 23811454
Abstract
Aerva javanica (Burm.f.) Juss. ex Schult. seed essential oils were obtained by hydrodistillation (HD) and dry steam distillation (SD) extracting methods and analyzed by using gas chromatography-mass spectrometry(GC-MS). Twenty and eighteen components representing 90.5% and 95.6% of the seed essential oil were identified, using hydrodistillation and dry steam distillation, respectively. The major constituent identified from seed essential oil obtained by HD were heptacosane (25.4%), 3-allyl-6-methoxyphenol (14.1%), pentacosane (12.1%), 6,10,14-trimethyl-2-pentade-canone (7.9%), nonacosane (7.1%), tricosane (3.6%), α-farnesene (3.5%), dodecanal (2.7%) and octacosane (2.1%). Whereas the major constituent identified from seed essential oil obtained by SD were heptacosane (41.4%), pentacosane (21.2%), nonacosane (14.8%), tricosane (6.3%), octacosane (4.2%) and tetracosane (3.0%).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
286931
Pentacosane, 99%
C25H52