EMAIL THIS PAGE TO A FRIEND
Arteriosclerosis, thrombosis, and vascular biology
Atrial natriuretic peptide-mediated inhibition of microcirculatory endothelial Ca2+ and permeability response to histamine involves cGMP-dependent protein kinase I and TRPC6 channels.
PMID 23814119
Abstract
Histamine increases microvascular endothelial leakage by activation of complex calcium-dependent and -independent signaling pathways. Atrial natriuretic peptide (ANP) via its cGMP-forming guanylyl cyclase-A (GC-A) receptor counteracts this response. Here, we characterized the molecular mechanisms underlying this interaction, especially the role of cGMP-dependent protein kinase I (cGKI). We combined intravital microscopy studies of the mouse cremaster microcirculation with experiments in cultured microvascular human dermal endothelial cells. In wild-type mice, ANP had no direct effect on the extravasation of fluorescent dextran from postcapillary venules, but strongly reduced the histamine-provoked vascular leakage. This anti-inflammatory effect of ANP was abolished in mice with endothelial-restricted inactivation of GC-A or cGKI. Histamine-induced increases in endothelial [Ca(2+)]i in vitro and of vascular leakage in vivo were markedly attenuated by the Ca(2+)-entry inhibitor SKF96365 and in mice with ablated transient receptor potential canonical (TRPC) 6 channels. Conversely, direct activation of TRPC6 with hyperforin replicated the hyperpermeability responses to histamine. ANP, via cGKI, stimulated the inhibitory phosphorylation of TRPC6 at position Thr69 and prevented the hyperpermeability responses to hyperforin. Moreover, inhibition of cGMP degradation by the phosphodiesterase 5 inhibitor sildenafil prevented the edematic actions of histamine in wild types but not in mice with endothelial GC-A or cGKI deletion. ANP attenuates the inflammatory actions of histamine via endothelial GC-A/cGMP/cGKI signaling and inhibitory phosphorylation of TRPC6 channels. The therapeutic potential of this novel regulatory pathway is indicated by the observation that sildenafil improves systemic endothelial barrier functions by enhancing the endothelial effects of endogenous ANP.