EMAIL THIS PAGE TO A FRIEND
American journal of respiratory and critical care medicine
Rationalizing use of fluoroquinolones and pyrazinamide in the battle against multidrug-resistant tuberculosis.
PMID 23815718