EMAIL THIS PAGE TO A FRIEND
Journal of inorganic biochemistry
Antiproliferative effects of palladium(II) complexes of 5-nitrosopyrimidines and interactions with the proteolytic regulatory enzymes of the renin-angiotensin system in tumoral brain cells.
PMID 23816624
Abstract
Seventeen new palladium(II) complexes of general formulaes PdCl2L, PdCl(LH-1)(solvent) and PdCl2(PPh3)2L containing pyrimidine ligands derived from 6-amino-5-nitrosouracil and violuric acid have been prepared and characterized by elemental analysis, IR and NMR ((1)H and (13)C) methods and, two of them, PdCl(DANUH-1)(CH3CN)]·½H2O and [PdCl(2MeOANUH-1)(CH3CN)] by X-ray single-crystal diffraction (DANU: 6-amino-1,3-dimethyl-5-nitrosouracil; 2MeOANU: 6-amino-2-methoxy-5-nitroso-3H-pyrimidin-4-one). The coordination environment around palladium is nearly square planar in the two compounds with different supramolecular arrangements. Crystallographic and spectral data are consistent with a bidentate coordination mode through N5 and O4 atoms when the ligands act in neutral form and N5 and N6 atoms in the monodeprotonated ones. The cytotoxicity of the complexes against human neuroblastoma (NB69) and human glioma (U373-MG) cell lines has been tested showing a considerable antiproliferative activity. Also, the study of the effects of palladium(II) complexes on the renin-angiotensin system (RAS) regulating proteolytic regulatory enzymes aminopeptidase A (APA), aminopeptidase N (APN) and insulin-regulated aminopeptidase (IRAP) shows a strong dependence on the compound tested and the tumoral cell type, also affecting different catalytic routes; the compounds affect in a different way the activities of enzymes of the RAS system, changing their functional roles as initiators of cell proliferation in tumors as autocrine/paracrine mediators.