EMAIL THIS PAGE TO A FRIEND
Nature methods
Heritable genome editing in C. elegans via a CRISPR-Cas9 system.
PMID 23817069
Abstract
We report the use of clustered, regularly interspaced, short palindromic repeats (CRISPR)-associated endonuclease Cas9 to target genomic sequences in the Caenorhabditis elegans germ line using single-guide RNAs that are expressed from a U6 small nuclear RNA promoter. Our results demonstrate that targeted, heritable genetic alterations can be achieved in C. elegans, providing a convenient and effective approach for generating loss-of-function mutants.