EMAIL THIS PAGE TO A FRIEND
Vestnik Rossiiskoi akademii meditsinskikh nauk
[Fundamental bases of search of medicines for therapy of a diabetes mellitus type 2].
PMID 23819328
Abstract
In the presented review the data on searching for new drugs for diabetes mellitus treantment are analyzed. These drugs are used for metabolic disorder correction leading to hyperglycemia: beta-cells dysfunction, peripheral insulin resistance, increased hepatic glucose output.