EMAIL THIS PAGE TO A FRIEND
The American journal of psychiatry
Harnessing N-methyl-d-aspartate receptors for new treatment development in psychiatry: positive lessons from negative studies.
PMID 23820828