EMAIL THIS PAGE TO A FRIEND
Doklady biological sciences : proceedings of the Academy of Sciences of the USSR, Biological sciences sections
Sodium nitrite influences metabolic conversions of nitric oxide in tissues of the right and left ventricles of the rat heart.
PMID 23821046