EMAIL THIS PAGE TO A FRIEND
The New England journal of medicine
False positive galactomannan test after ice-pop ingestion.
PMID 23822795
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
48230 D-Galacto-D-mannan from Ceratonia siliqua, ~95% (HPLC)