EMAIL THIS PAGE TO A FRIEND
Clinical and experimental medicine
Programmed death-1 (PD-1)-dependent functional impairment of CD4(+) T cells in recurrent genital papilloma.
PMID 23824147
Abstract
Genital papilloma is caused by human papilloma virus (HPV) infection and recurs frequently. Although T cells are known to play a critical role in the control of HPV infection and papilloma development, the function and phenotype of these cells in the lesion remain to be elucidated. In the present study, we examined the function and phenotype of CD4(+) T cells isolated from the lesions of primary (n = 9) and recurrent (n = 11) genital papillomas. In recurrent papillomas, the frequency of proliferating (Ki-67(+)) CD4(+) T cells was significantly reduced compared with primary papillomas. Cytokine production was evaluated by intracellular cytokine staining in anti-CD3/anti-CD28-stimulated CD4(+) T cells. CD4(+) T cells from recurrent lesions showed impaired production of IL-2, IFN-γ, and TNF-α. Of interest, the frequency of cytokine-producing CD4(+) T cells significantly correlated with the frequency of Ki-67(+)CD4(+) T cells. We also studied expression of programmed death-1 (PD-1), a T-cell exhaustion marker. The frequency of PD-1(+)CD4(+) T cells was significantly increased in recurrent lesions and inversely correlated with the frequency of cytokine-producing CD4(+) T cells. The functional significance of PD-1 expression was determined in blocking assays with anti-PD-L1, which restored cytokine production of CD4(+) T cells from recurrent lesions. Taken together, in recurrent genital papilloma lesions, proliferation, and cytokine production by CD4(+) T cells are impaired and the PD-1/PD-L1 interaction is responsible for the functional impairment of CD4(+) T cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2501381
Anti-Cd28 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2501674
Anti-CD28 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405583
Anti-CD28 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
C7831 Anti-CD28 antibody, Mouse monoclonal, clone CD28.2, purified from hybridoma cell culture
SAB1412683
ANTI-IFNA1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1410692
Anti-IFNA1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
GW22461F Anti-Interleukin-2 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
I8398 Anti-Interleukin-2 antibody produced in goat, IgG fraction of antiserum, lyophilized powder
I8273 Anti-Interleukin-2 antibody produced in goat, IgG fraction of antiserum, lyophilized powder
I7401 Anti-Interleukin-2 antibody produced in goat, affinity isolated antibody, lyophilized powder
I7784 Anti-Interleukin-2 antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB4700135 Monoclonal Anti-CD28 antibody produced in mouse, low endotoxin, clone CD28.2, purified immunoglobulin, buffered aqueous solution
SAB4700739 Monoclonal Anti-CD28 antibody produced in mouse, clone JJ319, purified immunoglobulin, buffered aqueous solution
SAB4700751
Monoclonal Anti-CD28 antibody produced in mouse, clone CD28.2, purified immunoglobulin, buffered aqueous solution
WH0003439M7
Monoclonal Anti-IFNA1 antibody produced in mouse, clone 3C6, purified immunoglobulin, buffered aqueous solution
WH0003558M3 Monoclonal Anti-IL2 antibody produced in mouse, clone 3C8, purified immunoglobulin, buffered aqueous solution
I6784 Monoclonal Anti-Interleukin-2 antibody produced in mouse, clone 5334, purified immunoglobulin, lyophilized powder
I6909 Monoclonal Anti-Interleukin-2 antibody produced in mouse, clone 5355, purified immunoglobulin, lyophilized powder