EMAIL THIS PAGE TO A FRIEND
Journal of clinical microbiology
Meningitis and endocarditis caused by Campylobacter fetus after raw-liver ingestion.
PMID 23824770
Abstract
We report Campylobacter fetus meningitis associated with endocarditis in a 75-year-old diabetic man after he consumed raw liver. C. fetus was isolated from blood samples and cerebrospinal fluid. Cure was obtained with combined intravenous imipenem-gentamicin for 4 weeks; no relapse occurred after 6 months of follow-up.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I0160
Imipenem monohydrate, ≥98% (HPLC)
C12H17N3O4S · H2O