EMAIL THIS PAGE TO A FRIEND
Scandinavian journal of infectious diseases
Daptomycin-induced acute eosinophilic pneumonia: analysis of the current data and illustrative case reports.
PMID 23826793
Abstract
Acute eosinophilic pneumonia (AEP) is a rare but important complication of daptomycin therapy. We describe 2 cases of daptomycin-associated AEP, compile available data from another 22 published cases, and propose a revised set of diagnostic criteria.