EMAIL THIS PAGE TO A FRIEND
Annals of the Royal College of Surgeons of England
Use of a povidone-iodine sponge to protect skin and prevent K-wire infection.
PMID 23827296
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PVP1
Poly(vinylpyrrolidone)–Iodine complex