EMAIL THIS PAGE TO A FRIEND
Nanoscale
Fluorescent silver nanoclusters as DNA probes.
PMID 23828021
Abstract
Fluorescent silver nanoclusters (few atoms, quantum sized) have attracted much attention as promising substitutes for conventional fluorophores. Due to their unique environmental sensitivities, new fluorescent probes have been developed based on silver nanoclusters for the sensitive and specific detection of DNA. In this review we present the recent discoveries of activatable and color-switchable properties of DNA-templated silver nanoclusters and discuss the strategies to use these new properties in DNA sensing.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
85131
Silver, colloidal, 65-75% Ag basis
Ag
373249
Silver, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.99% trace metals basis
Ag
327077
Silver, flakes, 10 μm, ≥99.9% trace metals basis
Ag
326976
Silver, foil, thickness 2.0 mm, 99.9% trace metals basis
Ag
348724
Silver, foil, thickness 1.5 mm, 99.9% trace metals basis
Ag
369438
Silver, foil, thickness 1.0 mm, 99.99% trace metals basis
Ag
265543
Silver, foil, thickness 1.0 mm, 99.9% trace metals basis
Ag
265535
Silver, foil, thickness 0.5 mm, 99.99% trace metals basis
Ag
345075
Silver, foil, thickness 0.5 mm, 99.9% trace metals basis
Ag
348716
Silver, foil, thickness 0.25 mm, 99.99% trace metals basis
Ag
326984
Silver, foil, thickness 0.25 mm, 99.9% trace metals basis
Ag
265527
Silver, foil, thickness 0.1 mm, 99.9% trace metals basis
Ag
326992
Silver, foil, thickness 0.075 mm, 99.9% trace metals basis
Ag
265519
Silver, foil, thickness 0.025 mm, 99.9% trace metals basis
Ag
303372
Silver, granular, >250 μm, 99.99% trace metals basis
Ag
461032
Silver, needles, 250-600 μm, ≥99.99% trace metals basis
Ag
265500
Silver, powder, <250 μm, 99.99% trace metals basis
Ag
327107
Silver, powder, <45 μm, ≥99.99% trace metals basis
Ag
327093
Silver, powder, 5-8 μm, ≥99.9% trace metals basis
Ag
327085
Silver, powder, 2-3.5 μm, ≥99.9% trace metals basis
Ag
484059
Silver, nanopowder, <150 nm particle size, 99% trace metals basis
Ag
265624
Silver, rod, diam. 7.0 mm, 99.99% trace metals basis
Ag
348775
Silver, rod, diam. 3.2 mm, 99.98% trace metals basis
Ag
204366
Silver, shot, 1-3 mm, 99.999% trace metals basis
Ag
327050
Silver, shot, 1-3 mm, ≥99.99% trace metals basis
Ag
267430
Silver, wire, diam. 2.0 mm, ≥99.99% trace metals basis
Ag
327018
Silver, wire, diam. 2.0 mm, 99.9% trace metals basis
Ag
348759
Silver, wire, diam. 1.5 mm, ≥99.99% trace metals basis
Ag
265594
Silver, wire, diam. 1.0 mm, ≥99.99% trace metals basis
Ag
265608
Silver, wire, diam. 1.0 mm, 99.9% trace metals basis
Ag