EMAIL THIS PAGE TO A FRIEND
The Journal of family practice
Polymyalgia rheumatica and giant cell arteritis: how best to approach these related diseases.
PMID 23828809
Abstract
Each of these painful disorders can be challenging to diagnose, and many patients have both. Adequate glucocorticoid dosing and gradual tapering are key to treatment success.