EMAIL THIS PAGE TO A FRIEND
Fiziolohichnyi zhurnal (Kiev, Ukraine : 1994)
[Pathogenetic substantiation of using peptidases in the treatment of patients with generalized periodontitis].
PMID 23828974
Abstract
76 patients of whom 52 patients with GP-II degree exacerbative and 24 somatically healthy people aged 20-50 years were studied. Based on these immunological examination serapeptydase was assigned per os, as a complementary product to the basic therapy. Any concentration of secretory leukocyte protease inhibitor (SLPI) indicates the dominant hyperergic inflammation in periodontal tissues, accompanied by increased proteolysis. Profitable accumulation in the serum of patients with GP inhibitors proteinase to 53%, IL-1beta - to 26.3%, IL-2 - to 19.5%, IL-8 - to 42.8% FNPalpha - up to 48.3%, and reduction of IFNalpha to 78.1% indicates changes of complex "protease-antiproteases" immunocompetent cells and activation of inflammatory processes at the local and systemic levels.