EMAIL THIS PAGE TO A FRIEND
Medycyna pracy
[Toxicity of acrylamide and its metabolite - glicydamide].
PMID 23829070
Abstract
Acrylamide is a synthetic chemical compound commonly used in many branches of industry. It is mainly used in the synthesis of polyacrylamides, which are widely employed in plastics, paints, varnishes, adhesives and mortars production. Acrylamide is also applied in the cellulose-paper and cosmetic industries to produce toiletries and cosmetics. The interest in acrylamide increased in 2002, when Swedish scientists showed that a considerable amount of this substance is formed during frying and baking of various foods. Studies concerning toxicity of acrylamide and its metabolite - glicydamide showed their neurotoxic, genotoxic and carcinogenic effects. Neverthless, in humans only neurotoxic effect of acrylamide has been clearly evidenced. Genotoxic nature of acetylamide manifests itself mainly in its metabolic conversion to the epoxide derivative glicydamide. Carcinogenic effects of acrylamide have been shown in animal studies. Epidemiological studies have not provided explicit evidence that acrylamide supplied with the diet can initiate the formation of tumors in humans. Acrylamide exposure is assessed by measuring specific compounds (adducts) formed during the reaction of acrylamide with hemoglobin and DNA.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32458
Glycarbylamide, PESTANAL®, analytical standard
C5H6N4O2