EMAIL THIS PAGE TO A FRIEND
Alzheimer's research & therapy
A survey of TREM2 antibodies reveals neuronal but not microglial staining in formalin-fixed paraffin-embedded postmortem Alzheimer's brain tissues.
PMID 23830013