EMAIL THIS PAGE TO A FRIEND
Bioresource technology
Microalgal cultivation in wastewater from the fermentation effluent in Riboflavin (B2) manufacturing for biodiesel production.
PMID 23831749
Abstract
In this work, the acclimation of Chlorella pyrenoidosa in diluted wastewater was studied to produce biomass and remove chemical oxygen demand (COD), ammonia-N and phosphorous. The results indicated that the optimal conditions (the volume ratio of wastewater, light intensity, culture temperature, CO2 concentration in feeding gas) which could influence the wastewater treatment efficiency were 0.05, 250 photons m(-2) s(-1), 28 °C and 5%, respectively. Under these conditions, the removal efficiency of COD reached up to 89.2%, while the total nitrogen and total phosphorous decreased by 64.52% and 82.20%, respectively. With the second treatment, COD in the wastewater was further reduced to less than 100 mg/L while it was only reduced to 542.9 mg/L after the first treatment. The treated wastewater could be discharged directly or subjected to for further treatment for recycling. In addition, 1.25 g/L of the biomass and 38.27% (dry basis, w%) of lipid content were reached after microalgal cultivation.