EMAIL THIS PAGE TO A FRIEND
Middle East journal of anaesthesiology
Analgesic efficacy of continuous intravenous magnesium infusion as an adjuvant to morphine for postoperative analgesia: a systematic review and meta-analysis.
PMID 23833845
Abstract
The efficacy of perioperative intravenous magnesium administration on postoperative opioid use, opioid-related side effects (e.g., nausea and vomiting) and pain are uncertain, as randomized controlled trials on this topic have reported disparate results. The objective of this systematic review is to determine if perioperative magnesium reduces opioid use, opioid-related side effects, and postoperative pain. An electronic search was conducted using the Library of Medicine's PubMed and EMBASE databases. Included studies consisted of randomized controlled trials in an adult population with a clearly defined comparison of perioperative intravenous magnesium administration to a control with a documented assessment of opioid usage and postoperative pain. Relevant data was abstracted from included studies. Pooled estimates for weighted mean difference (WMD) with 95% confidence intervals (CI) were obtained for our primary outcome (opioid usage) using the Cochrane Collaboration's RevMan version 4.2.7 (Cochrane Collaboration; Oxford, United Kingdom). WMD and odds ratios (OR) were calculated using a random effects model. The literature search ultimately yielded 22 trials, enrolling 1177 (599 magnesium, 578 control) patients, who were included in the analysis. A significant decrease in morphine usage by those patients who received magnesium was noted (WMD = -7.40; 95% CI: -9.40 to -5.41, p < 0.00001). Perioperative magnesium administration was not associated with a difference in postoperative nausea or vomiting (RR = 0.76; 95% CI: 0.52 to 1.09, p = 0.14). The pooled visual analog scores for pain at 4-6 hours after surgery were significantly less in those patients who received magnesium surgery (WMD = -0.67; 95% CI: -1.12 to -0.23, p = 0.003); however, there was no difference in pain scores at 20-24 hours after surgery (WMD = -0.25; 95% CI: -0.62 to 0.71, p = 0.17). Based on the results of this systematic review, perioperative intravenous magnesium may be a useful adjuvant for the management of postoperative pain providing analgesia through a different mechanism of action than that of opioids and would make a potential addition to a multimodal anlgesic treatment plan; however, the decrease in opioid use with perioperative magnesium infusion does not appear to be associated with a decresea in opioid-related side effects.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
254118
Magnesium, chips, 6-35 mesh, 99.98% trace metals basis
Mg
474754
Magnesium, dendritic pieces, purified by distillation, 99.998% trace metals basis
Mg
465992
Magnesium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Mg
254126
Magnesium, 20-230 mesh, reagent grade, 98%
Mg
63040
Magnesium, grit, ≥99.0% (KT)
Mg
253987
Magnesium, ReagentPlus®, powder, −50 mesh, ≥99%
Mg
13112
Magnesium, powder, ≥99%
Mg
13103
Magnesium, ribbon, ≥99.5% Mg basis
Mg
299405
Magnesium, rod, diam. 6 mm, ≥99.9% trace metals basis
Mg
403148
Magnesium, turnings, ~1/8 in., 99.95% trace metals basis
Mg
200905
Magnesium, turnings, reagent grade, 98%
Mg
63035
Magnesium, purum, for Grignard reactions, ≥99.5%, turnings
Mg
723312
Magnesium, in a Sure/Seal bottle, turnings, 37.5 mmol
Mg
723320
Magnesium, in a Sure/Seal bottle, turnings, anhydrous tetrahydrofuran 37.5 mmol
Mg
735779
Magnesium, ingot, ≥99% trace metals basis
Mg
734365
Magnesium, wire, 127 μm diameter, 99.9% trace metals basis
Mg
735760
Magnesium, plate, 99.95%
Mg
GF86050199
Magnesium, microfoil, 25x25mm, thinness 1.0μm, specific density 174μg/cm2, 6 micron aluminum permanent support, 99.9%
Mg
GF67975172
Magnesium, wire reel, 2m, diameter 1.1mm, as drawn, 99.9+%
Mg
GF80461964
Magnesium, microleaf, 25x25mm, thinness 1.0μm, specific density 174μg/cm2, removable support, 99.9%
Mg
GF57746166
Magnesium, microleaf, 50x50mm, thinness 1.0μm, specific density 174μg/cm2, removable support, 99.9%
Mg
GF18270637
Magnesium, wire reel, 0.1m, diameter 0.125mm, as drawn, 99.9+%
Mg
GF96495609
Magnesium, wire reel, 0.1m, diameter 0.25mm, as drawn, 99.9+%
Mg
GF04987547
Magnesium, wire reel, 0.1m, diameter 0.4mm, as drawn, 99.9+%
Mg
GF23637371
Magnesium, wire reel, 0.1m, diameter 0.5mm, as drawn, 99.9+%
Mg
GF83427745
Magnesium, wire reel, 0.1m, diameter 1.0mm, as drawn, 99.9+%
Mg
GF86178189
Magnesium, wire reel, 0.1m, diameter 1.1mm, as drawn, 99.9+%
Mg
GF83829178
Magnesium, wire reel, 0.5m, diameter 0.125mm, as drawn, 99.9+%
Mg
GF61950618
Magnesium, wire reel, 0.5m, diameter 0.25mm, as drawn, 99.9+%
Mg
GF23443646
Magnesium, wire reel, 0.5m, diameter 0.4mm, as drawn, 99.9+%
Mg